Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SPARC-like 1/SPARCL1 Antibody (390), mFluor Violet 450 SE, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289237MFV450
Description
SPARC-like 1/SPARCL1 Monoclonal antibody specifically detects SPARC-like 1/SPARCL1 in Human samples. It is validated for Immunocytochemistry/ ImmunofluorescenceSpecifications
| SPARC-like 1/SPARCL1 | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunocytochemistry | |
| 390 | |
| Immunocytochemistry/ Immunofluorescence | |
| hevin, High endothelial venule protein, MAST 9, MAST9, PIG33, proliferation-inducing protein 33, SC1, SPARC-like 1 (hevin), SPARC-like 1 (mast9, hevin), SPARC-like protein 1 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human SPARC-like 1/SPARCL1 (Accession#: NP_004675.3; Met1-Phe664). | |
| 0.1 mL | |
| Cancer | |
| 8404 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction