Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SPARC-like 1/SPARCL1 Antibody Pair (HRP), R&D Systems™
SDP

Catalog No. NB003009 Shop All Novus Products
Change view
Click to view available options
Quantity:
15 Plates
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB003009 15 Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB003009 Supplier R&D Systems Supplier No. NBP27884615PLATES
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. Mouse Monoclonal Antibody

SPARC-like 1/SPARCL1 Monoclonal specifically detects SPARC-like 1/SPARCL1 in Human samples. It is validated for ELISA/Sandwich ELISA.

Specifications

Antigen SPARC-like 1/SPARCL1
Applications ELISA, Sandwich ELISA
Classification Monoclonal
Conjugate HRP
Dilution Sandwich ELISA
Gene Alias hevin, High endothelial venule protein, MAST 9, MAST9, PIG33, proliferation-inducing protein 33, SC1, SPARC-like 1 (hevin), SPARC-like 1 (mast9, hevin), SPARC-like protein 1
Gene Symbols SPARCL1
Host Species Mouse
Quantity 15 Plates
Gene ID (Entrez) 8404
Test Specificity This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human SPARC-like 1/SPARCL1.
Target Species Human
Content And Storage Storage of components varies. See protocol for specific instructions.
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.