Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SPARC Mouse anti-Human, Clone: 1A2C1, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
Antigen | SPARC |
---|---|
Clone | 1A2C1 |
Concentration | 1 mg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-6853
![]() ![]() |
Proteintech Group Inc
664261IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
SPARC is a secreted protein, acidic and rich in cysteines. It is a matrix-associated protein that elicits changes in cell shape, inhibits cell-cycle progression, and influences the synthesis of extracellular matrix (ECM)Specifications
SPARC | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Basement membrane protein 40, BM 40, ON, Osteonectin, SPARC | |
SPARC | |
IgG2a | |
Protein A | |
Antibody |
1A2C1 | |
Western Blot | |
Unconjugated | |
Mouse | |
Human | |
P09486 | |
6678 | |
SPARC Fusion Protein Ag7390 | |
Primary | |
-20°C | |
SPARC |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title