Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SPARC Polyclonal Antibody
GREENER_CHOICE

Catalog No. 01675842
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-675-842 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-675-842 Supplier Invitrogen™ Supplier No. PA112862
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody may cross react with SPARCL1. A suggested positive control for this product is mouse heart lysate.

SPARC gene encodes a cysteine-rich acidic matrix-associated protein. The encoded protein is required for the collagen in bone to become calcified but is also involved in extracellular matrix synthesis and promotion of changes to cell shape. The gene product has been associated with tumor suppression but has also been correlated with metastasis based on changes to cell shape which can promote tumor cell invasion. Three transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SPARC
Applications ELISA, Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.08% sodium azide
Gene SPARC
Gene Accession No. P09486
Gene Alias Basement membrane protein 40; basement-membrane protein 40; BM40; BM-40; Cysteine rich protein; OI17; ON; Osteonectin; osteonectin precursor; Secreted acidic cystein rich glycoprotein; secreted acidic cysteine rich glycoprotein; Secreted acidic cystein-rich glycoprotein (osteonectin); secreted protein acidic and cysteine rich; secreted protein acidic and rich in cysteine; Secreted protein acidic and rich in cysteine (SPARC); secreted protein, acidic, cysteine-rich (osteonectin); secreted protein, acidic, cysteine-rich [osteonectin]; SPARC
Gene Symbols SPARC
Host Species Rabbit
Immunogen Synthetic peptide corresponding to human SPARC.
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6678
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.