Learn More
Invitrogen™ SPARCL1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA580063
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human U-87MG whole cell, human SHG-44 whole cell, human MDA-MB-231 whole cell, human K562 whole cell, rat C6 whole cell, mouse smooth muscle tissue. IHC: human colon cancer tissue, mouse brain tissue.
This is a secreted protein. Found in the extracellular matrix. It's highly expressed in lymph node, brain, heart, lung, skeletal muscle, ovary, small intestine, and colon, with lower levels in placenta, pancreas, testis, spleen, and thymus, and no expression in kidney, liver, and peripheral blood leukocytes.
Specifications
SPARCL1 | |
Polyclonal | |
Unconjugated | |
SPARCL1 | |
Ecm2; Extracellular matrix protein 2; hevin; High endothelial venule protein; MAST 9; MAST9; matrix glycoprotein Sc1; PIG33; proliferation-inducing protein 33; SC1; SPARC like 1; SPARCL1; SPARC-like 1; SPARC-like 1 (hevin); SPARC-like 1 (mast9, hevin); SPARC-like protein 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
13602, 25434, 8404 | |
-20°C | |
Lyophilized |
Immunohistochemistry (Paraffin), Western Blot | |
500 μg/mL | |
PBS with 4mg trehalose and 0.05mg sodium azide | |
P24054, P70663, Q14515 | |
SPARCL1 | |
A synthetic peptide corresponding to a sequence of human SPARCL1 (ITRFFEECDPNKDKHITLKEWGHCFGIKEEDIDENLLF). | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.