Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SPARCL1 Polyclonal Antibody, Invitrogen™

Catalog No. PIPA547408
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA547408 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA547408 Supplier Thermo Scientific Supplier No. PA547408

Goat Polyclonal Antibody

In direct ELISAs, approximately 5% cross-reactivity with recombinant human SPARC-like 1 is observed and less than 1% cross-reactivity with recombinant mouse SPARC is observed. Reconstitute at 0.2 mg/mL in sterile PBS.

SPARCL1 Polyclonal antibody specifically detects SPARCL1 in Mouse samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SPARCL1
Applications Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene SPARCL1
Gene Accession No. P70663
Gene Alias Ecm2; Extracellular matrix protein 2; hevin; High endothelial venule protein; MAST 9; MAST9; matrix glycoprotein Sc1; PIG33; proliferation-inducing protein 33; SC1; SPARC like 1; SPARCL1; SPARC-like 1; SPARC-like 1 (hevin); SPARC-like 1 (mast9, hevin); SPARC-like protein 1
Gene Symbols SPARCL1
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse SPARC-like 1/SPARCL1 Ile17-Phe650
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13602
Target Species Mouse
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.