Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SPARCL1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501987054
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-7054 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-7054 Supplier Bioss Supplier No. BS6110R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This is a secreted protein. Found in the extracellular matrix. It′s highly expressed in lymph node, brain, heart, lung, skeletal muscle, ovary, small intestine, and colon, with lower levels in placenta, pancreas, testis, spleen, and thymus, and no expression in kidney, liver, and peripheral blood leukocytes.

Specifications

Antigen SPARCL1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene SPARCL1
Gene Accession No. Q14515
Gene Alias Ecm2; Extracellular matrix protein 2; Hevin; High endothelial venule protein; MAST 9; MAST9; Matrix glycoprotein Sc1; PIG33; proliferation-inducing protein 33; Sc1; SPARC like 1; Sparcl1; SPARC-like 1; SPARC-like 1 (hevin); SPARC-like 1 (mast9, hevin); SPARC-like protein 1
Gene Symbols SPARCL1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human SPARCL1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8404
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.