Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SPARCL1 Recombinant Rabbit Monoclonal Antibody (390), FITC
GREENER_CHOICE

Catalog No. PIMA540977
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA540977 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA540977 Supplier Invitrogen™ Supplier No. MA540977
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

May be stored for twelve months without detectable loss of activity. Protect from prolonged exposure to light and do not freeze. Sodium azide is toxic to cells and should be disposed of properly so flush with large volumes of water during disposal.

This is a secreted protein. Found in the extracellular matrix. It's highly expressed in lymph node, brain, heart, lung, skeletal muscle, ovary, small intestine, and colon, with lower levels in placenta, pancreas, testis, spleen, and thymus, and no expression in kidney, liver, and peripheral blood leukocytes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SPARCL1
Applications Flow Cytometry
Classification Recombinant Monoclonal
Clone 390
Concentration 0.1 mg/mL
Conjugate FITC
Formulation PBS with 0.5% BSA and 0.03% ProClin 300
Gene SPARCL1
Gene Accession No. Q14515
Gene Alias Ecm2; Extracellular matrix protein 2; hevin; High endothelial venule protein; MAST 9; MAST9; matrix glycoprotein Sc1; PIG33; proliferation-inducing protein 33; SC1; SPARC like 1; SPARCL1; SPARC-like 1; SPARC-like 1 (hevin); SPARC-like 1 (mast9, hevin); SPARC-like protein 1
Gene Symbols SPARCL1
Host Species Rabbit
Immunogen Recombinant Human SPARCL1 protein.
Purification Method Protein A
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8404
Target Species Human
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.