Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SPARCL1 Recombinant Rabbit Monoclonal Antibody (390), PE

Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA540978
This item is not returnable.
View return policy
Description
May be stored for twelve months without detectable loss of activity. Protect from prolonged exposure to light and do not freeze. Sodium azide is toxic to cells and should be disposed of properly so flush with large volumes of water during disposal.
This is a secreted protein. Found in the extracellular matrix. It's highly expressed in lymph node, brain, heart, lung, skeletal muscle, ovary, small intestine, and colon, with lower levels in placenta, pancreas, testis, spleen, and thymus, and no expression in kidney, liver, and peripheral blood leukocytes.
Specifications
| SPARCL1 | |
| Recombinant Monoclonal | |
| 0.1 mg/mL | |
| PBS with 0.5% BSA and 0.03% ProClin 300 | |
| Q14515 | |
| SPARCL1 | |
| Recombinant Human SPARCL1 protein. | |
| 100 Tests | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Flow Cytometry | |
| 390 | |
| PE | |
| SPARCL1 | |
| Ecm2; Extracellular matrix protein 2; hevin; High endothelial venule protein; MAST 9; MAST9; matrix glycoprotein Sc1; PIG33; proliferation-inducing protein 33; SC1; SPARC like 1; SPARCL1; SPARC-like 1; SPARC-like 1 (hevin); SPARC-like 1 (mast9, hevin); SPARC-like protein 1 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 8404 | |
| 4°C, store in dark, DO NOT FREEZE! | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction