Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Spastin Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA544807
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA544807 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA544807 Supplier Invitrogen™ Supplier No. PA544807
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: RVLVMGATNR PQELDEAVLR RFIKRVYVSL PNEETRLLLL KNLLCKQGSP Sequence homology: Cow: 100%; Dog: 100%; Goat: 93%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%.

This gene encodes a member of the AAA (ATPases associated with a variety of cellular activities) protein family. Members of this protein family share an ATPase domain and have roles in diverse cellular processes including membrane trafficking, intracellular motility, organelle biogenesis, protein folding, and proteolysis. The encoded ATPase may be involved in the assembly or function of nuclear protein complexes. Two transcript variants encoding distinct isoforms have been identified for this gene. Other alternative splice variants have been described but their full length sequences have not been determined. Mutations associated with this gene cause the most frequent form of autosomal dominant spastic paraplegia 4.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Spastin
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene SPAST
Gene Accession No. Q9UBP0
Gene Alias ADPSP; ATPase; fidgetin-like 1; FSP2; HAMAP-Rule:MF_03021}; hypothetical protein zgc:85952; Kiaa1083; KIAA183; LOW QUALITY PROTEIN: spastin; mKIAA1083; si:ch73-233a22.3; SPAST; spastic paraplegia 4 (autosomal dominant; spastic paraplegia 4 (autosomal dominant; spastin); spastic paraplegia 4 homolog; spastic paraplegia 4 protein; Spastin; spastin {ECO:0000255; SPG4; zgc:85952
Gene Symbols SPAST
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human SPAST.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6683
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.