Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SPAT5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557664
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557664 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557664 Supplier Invitrogen™ Supplier No. PA557664
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SLELSLQLSQ LDLEDTQIPT SRSTPYKPID DRITNKASDV LLDVTQSPGD GSGLMLEEVT GLKCNFESAR EGNEQLT Highest antigen sequence identity to the following orthologs: Mouse - 65%, Rat - 65%.

This gene encodes a member of the ATPase associated with diverse activities family, whose members are defined by a highly conserved ATPase domain. Members of this family participate in diverse cellular processes that include membrane fusion, DNA replication, microtubule severing, and protein degradation. The protein encoded by this gene has a putative mitochondrial targeting sequence and has been proposed to function in maintenance of mitochondrial function and integrity during mouse spermatogenesis. Allelic variants in this gene have been associated with epilepsy, hearing loss, and cognitive disability syndrome. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SPAT5
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SPATA5
Gene Accession No. Q8NB90
Gene Alias 2510048F20Rik; AFG2; ATPase family gene 2 homolog; ATPase family protein 2 homolog; C78064; EHLMRS; Spaf; Spata5; spermatogenesis associated 5; spermatogenesis associated factor; spermatogenesis associated factor SPAF; spermatogenesis-associated factor protein; Spermatogenesis-associated protein 5
Gene Symbols SPATA5
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SPAT5. Recombinant protein control fragment (Product #RP-102718).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 166378
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.