Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SPATA1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA572722
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA572722 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA572722 Supplier Invitrogen™ Supplier No. PA572722
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SPATA1 Polyclonal Antibody for Western Blot, IHC (P), ELISA

SPATA1 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SPATA1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene SPATA1
Gene Accession No. Q5VX52, Q9D5R4
Gene Alias 4921536I21Rik; AV266943; SP-2; SPAP1; Spata1; spermatogenesis associated 1; spermatogenesis associated protein 1; spermatogenesis-associated protein 1; Sperm-specific protein SP-2
Gene Symbols SPATA1
Host Species Rabbit
Immunogen SPATA1 antibody for an 18 amino acid synthetic peptide near the carboxy terminus of human SPATA1. The immunogen is within amino acids 300 - 350 of SPATA1.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100505741, 70951
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.