Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SPATA12 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA569734
Description
This target displays homology in the following species: Human: 100%.
This gene is expressed primarily in testis and may play a role in testicular development and spermatogenesis. The encoded protein may be upregulated in response to ultraviolet-C radiation. [provided by RefSeq, Dec 2015].
Specifications
SPATA12 | |
Polyclonal | |
Unconjugated | |
SPATA12 | |
SPATA12; spermatogenesis associated 12; spermatogenesis-associated protein 12; Spermatogenesis-related protein 5; SRG5; testicular tissue protein Li 184 | |
Rabbit | |
Affinity chromatography | |
RUO | |
353324 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q7Z6I5 | |
SPATA12 | |
synthetic peptide directed towards the N terminal region of human SPATA12. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction