Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SPATA16 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA583394
Description
Immunogen sequence: LQALHMLPQT VDWSSFPPQQ YLLTLGFKNK DDGKFLEKIS SRKLPIFTEH KTPFGLTRED TVRQMETMGK RIL.
This gene encodes a testis-specific protein that belongs to the tetratricopeptide repeat-like superfamily. The encoded protein localizes to the Golgi apparatus and may play a role in spermatogenesis.
Specifications
SPATA16 | |
Polyclonal | |
Unconjugated | |
SPATA16 | |
4921511F01Rik; 4930503K02Rik; CDC68; FACT; FACTP140; FLJ10857; FLJ14010; NYD-SP12; RGD1563726; Spata16; spermatogenesis associated 16; spermatogenesis-associated protein 16; spermatogenesis-related protein; SPGF6; SUPT16H; testicular tissue protein Li 185; testis development protein NYD-SP12; testis-specific Golgi protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
83893 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9BXB7 | |
SPATA16 | |
Recombinant protein corresponding to Human SPATA16. Recombinant protein control fragment (Product #RP-96368). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction