Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SPATA17 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA543863
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA543863 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA543863 Supplier Invitrogen™ Supplier No. PA543863
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: NMFLPFSSYH KNEKYIPSMH LSSKYGPISY KEQFRSENPK KWICDKDFQT Sequence homology: Human: 100%; Rabbit: 91%.

SPATA17 contains 3 IQ domains. The function of the SPATA17 protein is not known.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SPATA17
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene SPATA17
Gene Accession No. Q96L03
Gene Alias 1700065F16Rik; 4930504I07Rik; 4930513F16Rik; IQ motif containing H; IQCH; MSRG11; MSRG-11; RGD1565282; RP11-144C20.1; SPATA17; spermatogenesis associated 17; spermatogenesis-associated protein 17; Spermatogenesis-related protein 11; Srg11
Gene Symbols SPATA17
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human SPATA17 (aa 290-339).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 128153
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.