Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SPATA17 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA556341
Description
Immunogen sequence: KEPDPWELQL QKAKPLTHRR PKVKQKDSTS LTDWLACTSA RSFPRSEILP PINRKQCQGP FRDITEVLEQ RYRPLEPTLR VAEPIDEL Highest antigen sequence identity to the following orthologs: Mouse - 71%, Rat - 73%.
SPATA17 contains 3 IQ domains. The function of the SPATA17 protein is not known.
Specifications
| SPATA17 | |
| Polyclonal | |
| Unconjugated | |
| SPATA17 | |
| 1700065F16Rik; 4930504I07Rik; 4930513F16Rik; IQ motif containing H; IQCH; MSRG11; MSRG-11; RGD1565282; RP11-144C20.1; SPATA17; spermatogenesis associated 17; spermatogenesis-associated protein 17; Spermatogenesis-related protein 11; Srg11 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 128153 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96L03 | |
| SPATA17 | |
| Recombinant protein corresponding to Human SPATA17. Recombinant protein control fragment (Product #RP-94411). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction