Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SPATA18 Antibody - BSA Free, Novus Biologicals™

Chicken Polyclonal Antibody
$196.10 - $499.50
Specifications
Antigen | SPATA18 |
---|---|
Concentration | 1 mg/mL |
Applications | Western Blot, ELISA |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
SPATA18 Polyclonal specifically detects SPATA18 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA.Specifications
SPATA18 | |
Western Blot, ELISA | |
Unconjugated | |
RUO | |
PBS with 0.02% Sodium Azide | |
132671 | |
Antibody was raised against a 19 amino acid synthetic peptide near the amino terminus of human SPATA18. The immunogen is located within amino acids 90 - 140 of SPATA18. Amino Acid Squence: CDTFDRERHKDPSPRDRDMQ | |
Primary | |
At least three isoforms of SPATA18 are known to exist; this antibody will detect the two longest isoforms. SPATA18 antibody is predicted to not cross-react with other SPATA family members. |
1 mg/mL | |
Polyclonal | |
Chicken | |
Human, Mouse, Rat | |
FLJ32906, Mieap, mitochondria-eating protein, spermatogenesis associated 18 homolog (rat), Spermatogenesis-associated protein 18, SPETEX1 | |
SPATA18 | |
IgY | |
Affinity Purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title