Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SPATA18 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA551260

Catalog No. PA551260


Only null left
Add to Cart

Description

Description

The antibody detects endogenous levels of total SPATA18 protein.

SPATA18 (spermatogenesis associated 18), also known as Mieap or SPETEX1, is a 538 amino acid protein that is thought to play a role in cell differentiation during spermatogenesis, particularly during development from late elongate spematids to mature spermatozoa. Localizing to cytoplasm, SPATA18 is encoded by a gene that maps to human chromosome 4q12.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

SPATA18
Polyclonal
Unconjugated
SPATA18
1700067I02Rik; Mieap; Mitochondria-eating protein; Spata18; Spermatid-expressing gene 1 protein; spermatid-expressing gene-1; spermatogenesis associated 18; spermatogenesis associated 18 homolog; spermatogenesis-associated protein 18; SPETEX1; Spetex-1; testis tissue sperm-binding protein Li 71n
Rabbit
Antigen affinity chromatography
RUO
132671
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
1.6 mg/mL
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Q8TC71
SPATA18
Synthetic peptide of human SPATA18.
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.