Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SPATA18 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557848
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557848 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557848 Supplier Invitrogen™ Supplier No. PA557848
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AENLKRLVSN ETLRTLQEKL DFWLKEYNTN TCDQNLNHCL ELIEQVAKVQ GQLFGILTAA AQEGGRNDGV ETIKSR Highest antigen sequence identity to the following orthologs: Mouse - 67%, Rat - 70%.

SPATA18 (spermatogenesis associated 18), also known as Mieap or SPETEX1, is a 538 amino acid protein that is thought to play a role in cell differentiation during spermatogenesis, particularly during development from late elongate spematids to mature spermatozoa. Localizing to cytoplasm, SPATA18 is encoded by a gene that maps to human chromosome 4q12.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SPATA18
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SPATA18
Gene Accession No. Q8TC71
Gene Alias 1700067I02Rik; Mieap; Mitochondria-eating protein; Spata18; Spermatid-expressing gene 1 protein; spermatid-expressing gene-1; spermatogenesis associated 18; spermatogenesis associated 18 homolog; spermatogenesis-associated protein 18; SPETEX1; Spetex-1; testis tissue sperm-binding protein Li 71n
Gene Symbols SPATA18
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SPATA18. Recombinant protein control fragment (Product #RP-96705).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 132671
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.