Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SPATA18 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA572739
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA572739 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA572739 Supplier Invitrogen™ Supplier No. PA572739
Add to Cart
Add to Cart

Chicken Polyclonal Antibody

SPATA18 Polyclonal Antibody for Western Blot, ELISA

SPATA18 (spermatogenesis associated 18), also known as Mieap or SPETEX1, is a 538 amino acid protein that is thought to play a role in cell differentiation during spermatogenesis, particularly during development from late elongate spematids to mature spermatozoa. Localizing to cytoplasm, SPATA18 is encoded by a gene that maps to human chromosome 4q12.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SPATA18
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene SPATA18
Gene Accession No. Q0P557, Q6AYL6, Q8TC71
Gene Alias 1700067I02Rik; Mieap; Mitochondria-eating protein; Spata18; Spermatid-expressing gene 1 protein; spermatid-expressing gene-1; spermatogenesis associated 18; spermatogenesis associated 18 homolog; spermatogenesis-associated protein 18; SPETEX1; Spetex-1; testis tissue sperm-binding protein Li 71n
Gene Symbols SPATA18
Host Species Chicken
Immunogen SPATA18 antibody for a 19 amino acid synthetic peptide near the amino terminus of human SPATA18. The immunogen is within amino acids 90 - 140 of SPATA18.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 132671, 289586, 73472
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgY
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.