Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SPATA19 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113068
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113068 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113068 Supplier Invitrogen™ Supplier No. PA5113068
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SPATA19 Polyclonal Antibody for Western Blot, ELISA

May have a role in spermiogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SPATA19
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene SPATA19
Gene Accession No. Q7Z5L4, Q9DAQ9
Gene Alias 1700001L23Rik; AV047838; cancer/testis antigen 132; CT132; Spas1; SPATA19; spergen1; Spergen-1; spermatogenesis associated 19; spermatogenesis-associated protein 19, mitochondrial; Spermatogenic cell-specific gene 1 protein; spermatogenic specific-gene1; testis secretory sperm-binding protein Li 243mP
Gene Symbols SPATA19
Host Species Rabbit
Immunogen Recombinant Human Spermatogenesis-associated protein 19, mitochondrial protein (25-167AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 219938, 75469
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.