Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SPATA2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP204071
Description
SPATA2 Polyclonal specifically detects SPATA2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
SPATA2 | |
Polyclonal | |
Immunohistochemistry 1:100-1:500, Immunohistochemistry-Paraffin 1:10-1:500 | |
NP_006029.1 | |
SPATA2 | |
The immunogen recognized by this antibody maps to a region between residue 250 and 300 of human spermatogenesis associated 2 using the numbering given in entry NP_006029.1 (GeneID 9825). | |
0.1 mL | |
Primary | |
Human | |
IgG |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
TBS and 0.1% BSA with 0.09% Sodium Azide | |
KIAA0757FLJ13167, PD1spermatogenesis associated PD1, spermatogenesis associated 2, spermatogenesis-associated protein 2, Spermatogenesis-associated protein PD1, tamo | |
Rabbit | |
Affinity Purified | |
RUO | |
9825 | |
Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction