Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SPATA2 Antibody (Z1P1F10*F12), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250304H
Description
SPATA2 Monoclonal specifically detects SPATA2 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry.Specifications
SPATA2 | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
SPATA2 | |
Peptide Sequence - KSTQLSHLVYR | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot, ELISA, Immunohistochemistry | |
Z1P1F10*F12 | |
Western Blot, ELISA, Immunohistochemistry | |
KIAA0757FLJ13167, PD1spermatogenesis associated PD1, spermatogenesis associated 2, spermatogenesis-associated protein 2, Spermatogenesis-associated protein PD1, tamo | |
Mouse | |
Protein A purified | |
RUO | |
9825 | |
Store at 4C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction