Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SPATA2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA561772
Description
Immunogen sequence: TYFSTQDDVD LYTDSEPRAT YRRQDALRPD VWLLRNDAHS LYHKRSPPAK ESALSKCQSC GLSCSSSLCQ RCDSLLTCPP AS Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 85%.
May have a role in the regulation of spermatogenesis.
Specifications
SPATA2 | |
Polyclonal | |
Unconjugated | |
SPATA2 | |
AI504642; KIAA0757; mKIAA0757; PD1; PPP1R145; protein phosphatase 1, regulatory subunit 145; SPATA2; spermatogenesis associated 2; spermatogenesis associated PD1; spermatogenesis-associated protein 2; spermatogenesis-associated protein PD1; tamo | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
9825 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9UM82 | |
SPATA2 | |
Recombinant protein corresponding to Human SPATA2. Recombinant protein control fragment (Product #RP-92311). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction