Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SPATA2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA565714
Description
Immunogen sequence: KDYYKPRVTKP SRSVDAYDSY WESRKPPLKA SLSLRKEPVA TDVGDDLKDE IIRPSPSLLT MASSPHGSPD VL Highest antigen sequence identity to the following orthologs - mouse 90%, rat 92%.
May have a role in the regulation of spermatogenesis.
Specifications
SPATA2 | |
Polyclonal | |
Unconjugated | |
SPATA2 | |
AI504642; KIAA0757; mKIAA0757; PD1; PPP1R145; protein phosphatase 1, regulatory subunit 145; SPATA2; spermatogenesis associated 2; spermatogenesis associated PD1; spermatogenesis-associated protein 2; spermatogenesis-associated protein PD1; tamo | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
9825 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9UM82 | |
SPATA2 | |
Recombinant Human SPATA2. Recombinant protein control fragment (Product #RP-106385). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction