Learn More
SPATA2 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A302493A

Description
The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections.
May have a role in the regulation of spermatogenesis.Specifications
SPATA2 | |
Polyclonal | |
Unconjugated | |
SPATA2 | |
KIAA0757, PD1, PPP1R145, protein phosphatase 1, regulatory subunit 145, spermatogenesis associated PD1, spermatogenesis-associated protein PD1, tamo | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
9825 | |
4° C | |
Liquid |
Immunohistochemistry, Immunoprecipitation, Western Blot | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
Q9UM82 | |
SPATA2 | |
Between 250 and 300 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.