Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SPATA20 Antibody, Alexa Fluor™ 350, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297798AF350
Description
SPATA20 Polyclonal specifically detects SPATA20 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| SPATA20 | |
| Polyclonal | |
| Immunohistochemistry, Immunohistochemistry-Paraffin | |
| DKFZp686H1839, FLJ21347, FLJ21969, MGC111032, sperm protein SSP411, spermatogenesis associated 20, spermatogenesis-associated protein 20, Sperm-specific protein 411, Ssp411, Tisp78, transcript increased in spermiogenesis 78 | |
| Produced in rabbits immunized with E. coli-derived Human SPATA20 fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Alexa Fluor 350 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| 64847 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction