Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SPATA20 Antibody, DyLight 488, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297798G
Description
SPATA20 Polyclonal specifically detects SPATA20 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
SPATA20 | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
DKFZp686H1839, FLJ21347, FLJ21969, MGC111032, sperm protein SSP411, spermatogenesis associated 20, spermatogenesis-associated protein 20, Sperm-specific protein 411, Ssp411, Tisp78, transcript increased in spermiogenesis 78 | |
Produced in rabbits immunized with E. coli-derived Human SPATA20 fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
DyLight 488 | |
50mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
64847 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction