Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SPATA20 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5110878
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5110878 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5110878 Supplier Invitrogen™ Supplier No. PA5110878
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SHPSSTPQRV PNRLIHEKSP YLLQHAYNPV DWYPWGQEAF DKARKENKPI FLSVGYSTCH WCHMMEEESF QNEEIGRLLS EDFVSVKV Highest antigen sequence indentity to the following orthologs: Mouse - 82%, Rat - 83%.

The protein may play a role in fertility regulation. It were significantly upregulated in individual bile samples from Cholangiocarcinoma patients by Western blotting and Immunohistochemistry, relative to normal tissues. It acts as a potential serum diagnostic biomarker for Cholangiocarcinoma.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SPATA20
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SPATA20
Gene Accession No. Q8TB22
Gene Alias epididymis secretory protein Li 98; HEL-S-98; Spata20; sperm protein SSP411; spermatogenesis associated 20; spermatogenesis-associated protein 20; sperm-specific protein 411; SSP411; Tisp78; transcript increased in spermiogenesis 78; transcript increased in spermiogenesis 78 protein
Gene Symbols SPATA20
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human SPATA20. Recombinant protein control fragment (Product #RP-93216).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64847
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.