Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SPATA21 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA572045
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA572045 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA572045 Supplier Invitrogen™ Supplier No. PA572045
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SPATA21 Polyclonal Antibody for Western Blot

SPATA21 is involved in the differentiation of haploid spermatids.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SPATA21
Applications Western Blot
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene SPATA21
Gene Accession No. Q7Z572
Gene Alias 4933414G08; SPATA21; spergen2; spergen-2; Spergen3; spergen-3; spermatogenesis associated 21; spermatogenesis-associated protein 21; Spermatogenic cell-specific gene 3 protein
Gene Symbols SPATA21
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 396-424 amino acids from the C-terminal region of human SPATA21.
Purification Method Antigen Affinity Chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 374955
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.