Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SPATA21 Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS17626R

Description
SPATA21 is involved in the differentiation of haploid spermatids (By similarity).Specifications
SPATA21 | |
Polyclonal | |
Unconjugated | |
SPATA21 | |
SPATA21; spergen2; spergen-2; spermatogenesis associated 21; Spermatogenesis-associated protein 21 | |
Rabbit | |
Protein A | |
RUO | |
374955 | |
-20°C | |
Liquid |
Western Blot | |
1 μg/mL | |
TBS with 1% BSA, 50% glycerol and 0.03% Proclin 300; pH | |
Q7Z572 | |
SPATA21 | |
KLH conjugated synthetic peptide derived from human SPATA21. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction