Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SPATA22 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$487.50

Specifications

Antigen SPATA22
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP155225
SDP
View Documents
Novus Biologicals
NBP155225
100 μL
Each of 1 for $487.50
Only null left
Add to Cart
 
Description

Description

SPATA22 Polyclonal specifically detects SPATA22 in Human samples. It is validated for Western Blot.
Specifications

Specifications

SPATA22
Polyclonal
Rabbit
Q8NHS9
84690
Synthetic peptides corresponding to SPATA22(spermatogenesis associated 22) The peptide sequence was selected from the N terminal of SPATA22. Peptide sequence MKRSLSENSARSTAGCLPVPLFNQKKRNRQPLTSNPLKDDSGISTPSDNY.
Primary
Western Blot
Unconjugated
RUO
NYDSP20, NYD-SP20, spermatogenesis associated 22, spermatogenesis-associated protein 22, testes development-related NYD-SP20, Testis development protein NYD-SP20
SPATA22
IgG
40 kDa
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.