Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SPATA22 Antibody (OTI6H9), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274309F
Description
SPATA22 Monoclonal specifically detects SPATA22 in Human samples. It is validated for Immunohistochemistry.Specifications
SPATA22 | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
SPATA22 | |
Full length recombinant protein of human SPATA22 (NP_115987) produced in E.coli. | |
0.1 mL | |
84690.0 | |
Store at 4°C in the dark. |
Immunohistochemistry | |
OTI6H9 | |
Immunohistochemistry | |
NYDSP20, NYD-SP20, spermatogenesis associated 22, spermatogenesis-associated protein 22, testes development-related NYD-SP20, Testis development protein NYD-SP20 | |
Mouse | |
Affinity Purified | |
Primary | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction