Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SPATA22 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100335
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100335 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100335 Supplier Invitrogen™ Supplier No. PA5100335
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total SPATA22.

SPATA22 is highly expressed in adult testis. The specific function of SPATA22 is not yet known.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SPATA22
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene SPATA22
Gene Accession No. Q5SV06, Q8NHS9
Gene Alias Gm882; NYDSP20; NYD-SP20; Repro42; RGD1565378; Sgn; Spata22; spermatogenesis associated 22; spermatogenesis-associated protein 22; testicular tissue protein Li 186; testis development protein NYD-SP20; Unknown (protein for MGC:134099)
Gene Symbols SPATA22
Host Species Rabbit
Immunogen A synthesized peptide derived from human SPATA22(Accession Q8NHS9), corresponding to amino acid residues E7-T57.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 380709, 84690
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.