Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SPATA25 Antibody (Z37P3H11*B2), Janelia Fluor™ 549, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250305JF549
Description
SPATA25 Monoclonal specifically detects SPATA25 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry.Specifications
SPATA25 | |
Monoclonal | |
Janelia Fluor 549 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
SPATA25 | |
Peptide Sequence - LVRSKRGQP | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot, ELISA, Immunohistochemistry | |
Z37P3H11*B2 | |
Western Blot, ELISA, Immunohistochemistry | |
C20orf165, Chromosome 20 Open Reading Frame 165, DJ337O18.8, Spermatogenesis Associated 25, Testis-Specific Gene 23 Protein, TSG23 | |
Mouse | |
Protein A purified | |
Developmental Biology | |
128497 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction