Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SPATA25 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA571413
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA571413 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA571413 Supplier Invitrogen™ Supplier No. PA571413
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SPATA25 Polyclonal Antibody for Western Blot

May play a role in spermatogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SPATA25
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene SPATA25
Gene Accession No. Q9BR10
Gene Alias 1700020C07Rik; C20orf165; dJ337O18.8; Spata25; spermatogenesis associated 25; spermatogenesis-associated protein 25; Testis-specific gene 23 protein; TSG23
Gene Symbols SPATA25
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human SPATA25.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 128497
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.