Learn More
Invitrogen™ SPATA25 Polyclonal Antibody

Description
Immunogen sequence: SSQPDICILTL AMMIAGIPTV PVPGVREEDL IWAAQAFMMA HPEPEGAVEG ARWEQAHAHT ASGKMPLVRS KRGQ Highest antigen sequence identity to the following orthologs - mouse 86%, rat 89%.
Specifications
Specifications
| Antigen | SPATA25 |
| Applications | Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | SPATA25 |
| Gene Accession No. | Q9BR10 |
| Gene Alias | 1700020C07Rik; C20orf165; dJ337O18.8; Spata25; spermatogenesis associated 25; spermatogenesis-associated protein 25; Testis-specific gene 23 protein; TSG23 |
| Gene Symbols | SPATA25 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.