Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SPATA2L Antibody (OTI3F6), Alexa Fluor™ 594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274310AF594
Description
SPATA2L Monoclonal specifically detects SPATA2L in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
SPATA2L | |
Monoclonal | |
Alexa Fluor 594 | |
50 mM sodium borate with 0.05% sodium azide | |
SPATA2L | |
Recombinant protein fragment corresponding to amino acids 179-424 of human SPATA2L (NP_689552) produced in E.coli. | |
Primary | |
Human, Mouse, Rat | |
IgG2a |
Western Blot | |
OTI3F6 | |
Western Blot | |
C16orf76, chromosome 16 open reading frame 76, MGC26885, SPATA2-like protein, spermatogenesis associated 2-like, spermatogenesis-associated protein 2-like protein, tamo | |
Mouse | |
0.1 mL | |
124044 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction