Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SPATA2L Monoclonal Antibody (OTI2B4), TrueMAB™, OriGene

Mouse Monoclonal Antibody
$516.29
Specifications
Antigen | SPATA2L |
---|---|
Clone | OTI2B4 |
Concentration | 1 mg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-167-9212
![]() ![]() |
OriGene
TA802370 |
100 μL |
Each of 1 for $516.29
|
|
|||||
Description
SPATA2L Monoclonal antibody specifically detects SPATA2L in Human samples. It is validated for Western BlotSpecifications
SPATA2L | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
Q8IUW3 | |
IgG1 | |
-20° C, Avoid Freeze/Thaw Cycles |
OTI2B4 | |
Western Blot | |
Unconjugated | |
Mouse | |
Human | |
124044 | |
Primary | |
SPATA2L |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title