Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SPATA2L Monoclonal Antibody (OTI3F6)
GREENER_CHOICE

Catalog No. PIMA526538
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA526538 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA526538 Supplier Invitrogen™ Supplier No. MA526538
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SPATA2L Monoclonal Antibody for Western Blot, IHC (P)

SPATA2L belongs to the SPATA2 family. The exact function of SPATA2L is not known. There are two different isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SPATA2L
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI3F6
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene SPATA2L
Gene Accession No. Q8IUW3
Gene Alias C16orf76; SPATA2L; SPATA2-like protein; spermatogenesis associated 2 like; spermatogenesis associated 2-like; spermatogenesis-associated protein 2-like protein; tamo
Gene Symbols SPATA2L
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 179-424 of SPATA2L produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 124044
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.