Learn More
SPATA2L Mouse anti-Human, Clone: OTI3E5, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF802364

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
SPATA2L belongs to the SPATA2 family. The exact function of SPATA2L is not known. There are two different isoforms.Specifications
SPATA2L | |
Monoclonal | |
Unconjugated | |
SPATA2L | |
C16orf76, tamo | |
Mouse | |
Affinity Chromatography | |
RUO | |
124044 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI3E5 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q8IUW3 | |
SPATA2L | |
Human recombit protein fragment corresponding to amino acids 179-424 of human SPATA2L produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.