Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SPATA2L Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559981
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559981 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559981 Supplier Invitrogen™ Supplier No. PA559981
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VLKGVLSDDL LLKSFQKMGY VRRDSHRLMV TALPPACQLV QVALGCFALR LECEILGEVL AQLGTSVLPA EELLQARRAS GDVASCVAWL QQRLAQDE Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 91%.

SPATA2L belongs to the SPATA2 family. The exact function of SPATA2L is not known. There are two different isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SPATA2L
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SPATA2L
Gene Accession No. Q8IUW3
Gene Alias C16orf76; SPATA2L; SPATA2-like protein; spermatogenesis associated 2 like; spermatogenesis associated 2-like; spermatogenesis-associated protein 2-like protein; tamo
Gene Symbols SPATA2L
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SPATA2L. Recombinant protein control fragment (Product #RP-98572).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 124044
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.