Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SPATA2L Monoclonal Antibody (OTI3H8)
SPATA2L Monoclonal Antibody (OTI3H8)
Supplier: Thermo Scientific MA526537
Description

Specifications
SPATA2L | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8IUW3 | |
SPATA2L | |
Human recombinant protein fragment corresponding to amino acids 179-424 of SPATA2L produced in E.coli. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG2b |
Immunohistochemistry (Paraffin), Western Blot | |
OTI3H8 | |
Unconjugated | |
SPATA2L | |
C16orf76; SPATA2L; SPATA2-like protein; spermatogenesis associated 2 like; spermatogenesis associated 2-like; spermatogenesis-associated protein 2-like protein; tamo | |
Mouse | |
Affinity Chromatography | |
RUO | |
124044 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction