Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SPATA3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553765
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553765 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553765 Supplier Invitrogen™ Supplier No. PA553765
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MKKVKKKRSE ARRHRDSTSQ HASSNSTSQQ PSPESTPQQP SPESTPQHSS LETTSRQPAF QALPAPEIRR SSCCLLSPDA N Highest antigen sequence identity to the following orthologs: Mouse - 40%, Rat - 41%.

SPATA3 (Spermatogenesis Associated 3) is a Protein Coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SPATA3
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SPATA3
Gene Accession No. Q8NHX4
Gene Alias 1700011N12Rik; 1700029H01Rik; 4930424D10Rik; Mtsarg1; SPATA3; spermatogenesis associated 3; spermatogenesis-associated protein 3; testis and spermatogenesis cell apoptosis related gene 1; testis and spermatogenesis cell apoptosis related protein 1; testis and spermatogenesis cell-related protein 1; Testis spermatocyte apoptosis-related protein 1; Tsarg1
Gene Symbols SPATA3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SPATA3. Recombinant protein control fragment (Product #RP-92084).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 130560
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.