Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SPATA3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA572723
Description
SPATA3 Polyclonal Antibody for Western Blot, IHC (P), ELISA
SPATA3 (Spermatogenesis Associated 3) is a Protein Coding gene.
Specifications
SPATA3 | |
Polyclonal | |
Unconjugated | |
SPATA3 | |
1700011N12Rik; 1700029H01Rik; 4930424D10Rik; Mtsarg1; SPATA3; spermatogenesis associated 3; spermatogenesis-associated protein 3; testis and spermatogenesis cell apoptosis related gene 1; testis and spermatogenesis cell apoptosis related protein 1; testis and spermatogenesis cell-related protein 1; Testis spermatocyte apoptosis-related protein 1; Tsarg1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
130560, 70060 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
Q8NHX4, Q9D9T6 | |
SPATA3 | |
SPATA3 antibody for a 17 amino acid synthetic peptide near the carboxy terminus of human SPATA3. The immunogen is within the last 50 amino acids of SPATA3. | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction