Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SPATA31A2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA571097
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA571097 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA571097 Supplier Invitrogen™ Supplier No. PA571097
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Human: 100%.

SPATA31A1 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SPATA31A2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene SPATA31A1
Gene Accession No. Q5TZJ5
Gene Alias C9orf36; C9orf36A; FAM75A1; FAM75A2; family with sequence similarity 75, member A1; family with sequence similarity 75, member A2; protein FAM75A1; protein FAM75A2; SPATA31 subfamily A member 1; SPATA31 subfamily A, member 1; SPATA31 subfamily A, member 2; SPATA31A1; SPATA31A2; spermatogenesis-associated protein 31A1; spermatogenesis-associated protein 31A2
Gene Symbols SPATA31A1
Host Species Rabbit
Immunogen synthetic peptide directed towards the N-terminal region of human SPATA31A2.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 647060
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.