Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SPATA31D1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA561618
Description
Immunogen sequence: IRAAFTGTTE AQKIRKDTRE FLEEKLGHRH GIDITCPQEP LSFPVGLGKA QHNPEVHVRA EPVQGCPCNY RAPSCKVTRT KSCSQQAIFV GQNYPTRIRQ IIDKDRQPQK VEAFKGKILC QSHPQSMPHR KPVPHPNPTC R Highest antigen sequence identity to the following orthologs: Mouse - 31%, Rat - 26%.

Specifications
SPATA31D1 | |
Polyclonal | |
Unconjugated | |
SPATA31D1 | |
FAM75D1; FAM75-like protein FLJ46321; family with sequence similarity 75, member D1; protein FAM75D1; SPATA31 subfamily D member 1; SPATA31 subfamily D, member 1; SPATA31D1; Spermatogenesis-associated protein 31D1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
389763 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.6 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6ZQQ2 | |
SPATA31D1 | |
Recombinant protein corresponding to Human SPATA31D1. Recombinant protein control fragment (Product #RP-90571). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction