Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SPATA31E1 Antibody, Alexa Fluor™ 750, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP306105AF750
Description
SPATA31E1 Polyclonal specifically detects SPATA31E1 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
SPATA31E1 | |
Polyclonal | |
Immunocytochemistry/Immunofluorescence | |
C9orf79, chromosome 9 open reading frame 79, FAM75E1, FAM75-like protein C9orf79, SPATA31 subfamily E, member 1, XXyac-YM21GA2.5 | |
Produced in rabbits immunized with E. coli-derived Human SPATA31E1 fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunofluorescence | |
Alexa Fluor 750 | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
286234 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction