Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SPATA32 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA555104
Description
Immunogen sequence: SVHSNMGLPT PQTFRPWSLN SNCRSFTEEN HVSACHHSIS AQTSKHLFWA NKLIQASEHS LQRAINMQLN NGSAGQPIRS PL Highest antigen sequence identity to the following orthologs: Mouse - 37%, Rat - 39%.
C17orf46, also named as SPATA32 and TEX34, mainly expressed in Testis. It interacts with syntaxin-1 and ACTB. It's about 42-52 kDa in our detection.
Specifications
SPATA32 | |
Polyclonal | |
Unconjugated | |
SPATA32 | |
acrosome expressed 2; AEP2; C17orf46; CTD-2020K17.4; SPATA32; spermatogenesis associated 32; spermatogenesis-associated protein 32; testis expressed 34; Testis-expressed protein 34; testis-expressed sequence 34 protein; TEX34; VAD1.2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
124783 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96LK8 | |
SPATA32 | |
Recombinant protein corresponding to Human SPATA32. Recombinant protein control fragment (Product #RP-93504). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction