Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SPATA4 Antibody, Novus Biologicals™
SDP

Catalog No. NBP220467 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP220467 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP220467 Supplier Novus Biologicals Supplier No. NBP220467
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SPATA4 Polyclonal specifically detects SPATA4 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen SPATA4
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:5000-1:20000, Immunohistochemistry 10 - 1:500, Immunohistochemistry-Paraffin 1:100-1:1000
Formulation PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal
Gene Accession No. Q8NEY3
Gene Alias SPEF1B, spermatogenesis associated 4, spermatogenesis-associated protein 4, testis and spermatogenesis cell related protein 2, Testis and spermatogenesis cell-related protein 2, Testis spermatocyte apoptosis-related gene 2 protein, TSARG2MGC33432
Gene Symbols SPATA4
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human TSARG2. The exact sequence is proprietary.
Molecular Weight of Antigen 35 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 132851
Test Specificity Human, Mouse. Chimpanzee (100%).
Target Species Human, Mouse
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.